Learn More
Cayman Chemical MTP 131 (acetate), Purity: ≥95%, CAS: 1334953-95-5, 10 mg
SDP
Catalog No. 502950749
Encompass_Preferred
A mitochondria-targeted peptide antioxidant; reduces tert-butyl hydroperoxide-induced lipid peroxidation and apoptosis in SH-SY5Y cells from 0.001 to 1 nM; reduces infarct volume, hemispheric swelling, and GSH depletion in a mouse model of MCAO-induced acute cerebral ischemia at 2 mg/kg; increases survival, improves motor function, and decreases degeneration of the lumbar spinal cord in the SOD1G93A transgenic mouse model of ALS at 5 mg/kg; reduces albuminuria, urinary hydrogen peroxide levels, and mesangial matrix accumulation, as well as preserves superoxide production, in a db/db mouse model of diabetic nephropathy
Catalog No. 50-295-0749 Supplier Cayman Chemical Supplier No. 3330210 MG
A mitochondria-targeted peptide antioxidant; reduces tert-butyl hydroperoxide-induced lipid peroxidation and apoptosis in SH-SY5Y cells from 0.001 to 1 nM; reduces infarct volume, hemispheric swelling, and GSH depletion in a mouse model of MCAO-induced acute cerebral ischemia at 2 mg/kg; increases survival, improves motor function, and decreases degeneration of the lumbar spinal cord in the SOD1G93A transgenic mouse model of ALS at 5 mg/kg; reduces albuminuria, urinary hydrogen peroxide levels, and mesangial matrix accumulation, as well as preserves superoxide production, in a db/db mouse model of diabetic nephropathy