Learn More
Bruker Daltonics MTP SMALLANCHOR 384 BC
Catalog No. NC1349382
Encompass
MTP SmallAnchor 384 BC MTP SmallAnchor 384 BC: MALDI target for scout MTP ion sources with 384 SmallAnchors (approx. 400 μm diameter) for sample preparation plus 96 CalibAnchors (additional anchor for external calibration). - frame (#8074115) required - dedicated for autoflex and ultraflex series mass spectrometers - compatible with Dual ESI/MALDI Source
Catalog No. NC1349382 Supplier Bruker Daltonics Supplier No. 8280792
MTP SmallAnchor 384 BC MTP SmallAnchor 384 BC: MALDI target for scout MTP ion sources with 384 SmallAnchors (approx. 400 μm diameter) for sample preparation plus 96 CalibAnchors (additional anchor for external calibration). - frame (#8074115) required - dedicated for autoflex and ultraflex series mass spectrometers - compatible with Dual ESI/MALDI Source