Learn More
Cole-Parmer Cole-Parmer Five-Position Stirrer, 120 VAC
Supplier:  Cole-Parmer 5145021
Encompass
These multiposition stirrers offer more versatility than single-position stirrers. Stirrers are available with five or nine stirring positions. Each stirring position is individually controlled so you can perform different mixing applications simultaneously.
Catalog No. NC9089393