Learn More
Sigma Aldrich Fine Chemicals Biosciences MTPTM Taq DNA Polymerase T
Catalog No. 502907268
Encompass
MTP Taq DNA Polymerase is a recombinant thermostable enzyme from Thermus aquaticus expressed in E. coli and purified using a proprietary process to minimize levels of contaminating DNA. The enzyme has both 5-3 DNA polymerase and exonuclease activities is 95 kDa by SDS-PAGE and has no detectable endonuclease or 3-5 exonuclease activities. Each lot of MTP Taq undergoes strict quality control testing to ensure the absence of detectable levels of contaminating DNA.Contaminating DNA present in most other polymerase preparations often preclude or obscure the accurate interpretation of results especially when targeting conserved sequences e.g. bacterial 16S rRNA region. Through Sigmas proprietary DNA removal methods and strict quality control standards we can ensure the absence of the most commonly found contaminant DNA. Each lot of MTP Taq is assayed using PCR and primers specific to (1) the conserved region of bacterial 16S rRNA (2) the Taq expression vector and (3) the human bet
Catalog No. 50-290-7268 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. D74421500UN
MTP Taq DNA Polymerase is a recombinant thermostable enzyme from Thermus aquaticus expressed in E. coli and purified using a proprietary process to minimize levels of contaminating DNA. The enzyme has both 5-3 DNA polymerase and exonuclease activities is 95 kDa by SDS-PAGE and has no detectable endonuclease or 3-5 exonuclease activities. Each lot of MTP Taq undergoes strict quality control testing to ensure the absence of detectable levels of contaminating DNA.Contaminating DNA present in most other polymerase preparations often preclude or obscure the accurate interpretation of results especially when targeting conserved sequences e.g. bacterial 16S rRNA region. Through Sigmas proprietary DNA removal methods and strict quality control standards we can ensure the absence of the most commonly found contaminant DNA. Each lot of MTP Taq is assayed using PCR and primers specific to (1) the conserved region of bacterial 16S rRNA (2) the Taq expression vector and (3) the human bet