Learn More
Medchemexpress LLC MTR-106 | 1639357-93-9 | 99.4% | 525.62 | 25 MG
SDP
Catalog No. 5000363316
Encompass_Preferred
MTR-106 is a potent and orally active G-quadruplex stabilizer and RNA polymerase I inhibitor, primarily utilized in cancer research. It effectively induces apoptosis and inhibits cell growth in various cancer models.
  • Potent G-quadruplex stabilizer
  • RNA polymerase I inhibitor
  • Induces apoptosis
  • Inhibits cell growth
  • Shows antitumor activity in HR-deficient and PARPi-resistant cells
  • Suppresses tumor growth in BRCA-deficient and PARPi-resistant xenografts
Catalog No. 50-003-63316 Supplier Medchemexpress LLC Supplier No. 25MGHY148953
MTR-106 is a potent and orally active G-quadruplex stabilizer and RNA polymerase I inhibitor, primarily utilized in cancer research. It effectively induces apoptosis and inhibits cell growth in various cancer models.
  • Potent G-quadruplex stabilizer
  • RNA polymerase I inhibitor
  • Induces apoptosis
  • Inhibits cell growth
  • Shows antitumor activity in HR-deficient and PARPi-resistant cells
  • Suppresses tumor growth in BRCA-deficient and PARPi-resistant xenografts