Learn More
Cole-Parmer Oakton pH 6+ Handheld Meter with pH Probe and NIST-Traceable Calibration
Catalog No. 501961762 Shop All Cole-Parmer Products
Encompass
These meters are perfect for fast, accurate results. Automatic temperature compensation (ATC)ensures accuracy from 0 to 100C. Meters include a protective rubber boot that also acts as a meter stand.
Catalog No. 50-196-1762 Supplier Cole-Parmer Supplier No. 3561342
These meters are perfect for fast, accurate results. Automatic temperature compensation (ATC)ensures accuracy from 0 to 100C. Meters include a protective rubber boot that also acts as a meter stand.