Learn More
MSC Proto Micrometer Torque Wrench, 1/2 in, 70-350 Nm, 1 ea
Catalog No. 501920510
Encompass
This micrometer torque wrench features a 1/2 in drive and a ratcheting head, designed for industrial torque applications. It measures torque in Newton Meters across a range of 70 to 350 Nm, calibrated for accuracy in both clockwise and counter-clockwise directions.
  • Calibrated to +/- 4% clockwise and +/- 6% counter-clockwise at 20% to 100% of full scale
  • Calibration accomplished by manual loading
  • Suitable for general industry, heavy equipment, and maintenance needs
  • Made in USA
  • Graduation of 1.0 Nm
Catalog No. 50-192-0510 Supplier MSC Supplier No. 38362166
This micrometer torque wrench features a 1/2 in drive and a ratcheting head, designed for industrial torque applications. It measures torque in Newton Meters across a range of 70 to 350 Nm, calibrated for accuracy in both clockwise and counter-clockwise directions.
  • Calibrated to +/- 4% clockwise and +/- 6% counter-clockwise at 20% to 100% of full scale
  • Calibration accomplished by manual loading
  • Suitable for general industry, heavy equipment, and maintenance needs
  • Made in USA
  • Graduation of 1.0 Nm