Learn More
Aviva Systems Biology Corp MTRF1L Antibody - N-terminal region (ARP56319_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56319_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MTRF1L. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RF1ML. Alternate names: MRF1L, HMRF1L, mtRF1a, ApplicationWB. Species reactivity: Human
Catalog No. 50-014-61807