Learn More
Thermo Scientific MTRX 384DEPWL PP BLCK NS 80/CS
Catalog No. 50823861 Shop All Thermo Scientific Products
Encompass
Matrix Technologies ScreenMates 384 Deep Well Block, Round bottom, Square wells. Polypropylene, Non-Sterile. Clear. 80 plates per case.
Catalog No. 50-823-861 Supplier Thermo Scientific Supplier No. 4325
Add to Cart
Add to Cart
Matrix Technologies ScreenMates 384 Deep Well Block, Round bottom, Square wells. Polypropylene, Non-Sterile. Clear. 80 plates per case.