Learn More
Thermo Scientific Thermo Scientific™ Matrix 384 Deep Well Polypropylene Block
Catalog No. 50823862 Shop All Thermo Scientific Products
Encompass
Matrix Technologies ScreenMates 384 Deep Well Block, Round bottom, Square wells. Polypropylene, Sterile. Clear. 80 plates per case.
Catalog No. 50-823-862 Supplier Thermo Scientific Supplier No. 432611
Matrix Technologies ScreenMates 384 Deep Well Block, Round bottom, Square wells. Polypropylene, Sterile. Clear. 80 plates per case.