Learn More
Thermo Scientific Thermo Scientific MTRX BLK384VBTM PP PLTNS 80/CS
Catalog No. 50823637 Shop All Thermo Scientific Products
Encompass
Matrix Technologies ScreenMates Black 384 Well V-Bottom Polypropylene Microplates. Non-Sterile. Suitable for low-binding assays, sample storage, low volume liquid handling and use as a dilution plate. 80 plates per case.
Catalog No. 50-823-637 Supplier Thermo Scientific Supplier No. 4308
Matrix Technologies ScreenMates Black 384 Well V-Bottom Polypropylene Microplates. Non-Sterile. Suitable for low-binding assays, sample storage, low volume liquid handling and use as a dilution plate. 80 plates per case.