Learn More
Thermo Scientific Thermo Scientific Matrix Microplate
Catalog No. 50823638 Shop All Thermo Scientific Products
Encompass
Matrix Technologies ScreenMates Clear 384 Well V-Bottom Polypropylene Microplates. Sterile. Suitable for low-binding assays, sample storage, low volume liquid handling and use as a dilution plate. 80 plates per case.
Catalog No. 50-823-638 Supplier Thermo Scientific Supplier No. 4309
Add to Cart
Add to Cart
Matrix Technologies ScreenMates Clear 384 Well V-Bottom Polypropylene Microplates. Sterile. Suitable for low-binding assays, sample storage, low volume liquid handling and use as a dilution plate. 80 plates per case.