Learn More
Apexbio Technology LLC MTS Cell Proliferation Assay Kit(Synonyms: MTS Assay, CellTiter 96 AQueous Assay, MTS Cell Viability Assay), 500T.
SDP
Catalog No. 5000061348
Encompass_Preferred
The MTS Cell Proliferation Assay Kit employs MTS a tetrazolium salt compound as a reagent to quantify cellular proliferation and cytotoxicity Within metabolically active cells mitochondrial succinate dehydrogenase reduces MTS to a water-soluble formazan product detectable at 490 nm optical absorption The intensity measured correlates directly with cell viability and proliferation Compared with the traditional MTT assay this method simplifies procedures by removing the need for additional dissolution steps or medium changes minimizing potential assay artifacts Thus it is broadly applied in biomedical research involving cell viability testing drug screening and cytotoxicity assays
Catalog No. 50-000-61348 Supplier Apexbio Technology LLC Supplier No. K2250500
The MTS Cell Proliferation Assay Kit employs MTS a tetrazolium salt compound as a reagent to quantify cellular proliferation and cytotoxicity Within metabolically active cells mitochondrial succinate dehydrogenase reduces MTS to a water-soluble formazan product detectable at 490 nm optical absorption The intensity measured correlates directly with cell viability and proliferation Compared with the traditional MTT assay this method simplifies procedures by removing the need for additional dissolution steps or medium changes minimizing potential assay artifacts Thus it is broadly applied in biomedical research involving cell viability testing drug screening and cytotoxicity assays