Learn More
TRIBIOSCIENCE INC MTS Cell Viability Assay, 2000 Tests
SDP
Catalog No. NC3720217
Encompass
This MTS Cell Viability Assay Kit is a colorimetric method for sensitive quantification of viable cells in proliferation and cytotoxicity assay. The assay can be used for the measurement of cell proliferation in response to growth factors, cytokines, mitogens, and nutrients, etc. It can also be used for the analysis of cytotoxic compounds like anticancer drugs and many other toxic agents and pharmaceutical compounds.
Catalog No. NC3720217 Supplier TRIBIOSCIENCE INC Supplier No. TBS2004
This MTS Cell Viability Assay Kit is a colorimetric method for sensitive quantification of viable cells in proliferation and cytotoxicity assay. The assay can be used for the measurement of cell proliferation in response to growth factors, cytokines, mitogens, and nutrients, etc. It can also be used for the analysis of cytotoxic compounds like anticancer drugs and many other toxic agents and pharmaceutical compounds.