Learn More
Abcam MTS Reagent, cell proliferation assay reagent, 250MG
Catalog No. 502215399
Encompass_Preferred
MTS reagent for cell proliferation assay. Tetrazolium inner salt is used for determining cell viability in proliferation assays. Reduced in metabolically active cells to a colored formazan product that is soluble in cell culture media. This conversion is thought to be carried out by NAD(P)H-dependent dehydrogenase enzymes. - Purity > 98% - Soluble in DPBS to 2 mg/ml
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-221-5399 Supplier Abcam Supplier No. AB223881250MG
MTS reagent for cell proliferation assay. Tetrazolium inner salt is used for determining cell viability in proliferation assays. Reduced in metabolically active cells to a colored formazan product that is soluble in cell culture media. This conversion is thought to be carried out by NAD(P)H-dependent dehydrogenase enzymes. - Purity > 98% - Soluble in DPBS to 2 mg/ml
The product is subject to the following: Abcam Restricted Use Statement