Learn More
Cayman Chemical MTSEA-bIotIn 10mg
SDP
Catalog No. 502980359
Encompass_Preferred
A thiol-reactive probe that can be used to label proteins at cysteine residues
Catalog No. 50-298-0359 Supplier Cayman Chemical Supplier No. 1723710 MG
A thiol-reactive probe that can be used to label proteins at cysteine residues