Learn More
Cayman Chemical MTSEA-biotin, Purity: ≥95%, CAS: 162758-04-5, 5 mg
SDP
Catalog No. 502967136
Encompass_Preferred
A thiol-reactive probe that can be used to label proteins at cysteine residues
Catalog No. 50-296-7136 Supplier Cayman Chemical Supplier No. 172375 MG
A thiol-reactive probe that can be used to label proteins at cysteine residues