Learn More
Biotium Biotin-XX MTSEA,10mg
Catalog No. 501964817
Encompass_Preferred
MTSEA biotin selectively labels thiols under mild conditions. We offer MTSEA biotin with different linker lengths (Biotin-MTSEA, Biotin-X-MTSEA, and Biotin-XX-MTSEA), all of which allow selective labeling of cell surface thiols. Longer linkers should facilitate interaction between biotin and streptavidin. MTSEA biotin-XX MTSEA Biotincapcap; 2-((6-((6-((biotinoyl)amino)hexanoyl)amino)hexanoyl)amino)ethylmethanethiosulfonate Properties: White solid soluble in DMF or DMSO; Store desiccated at -20¿C; C25H45N5O6S3; MW: 607.7
Catalog No. 50-196-4817 Supplier Biotium Supplier No. 90066
Add to Cart
Edge
Add to Cart
MTSEA biotin selectively labels thiols under mild conditions. We offer MTSEA biotin with different linker lengths (Biotin-MTSEA, Biotin-X-MTSEA, and Biotin-XX-MTSEA), all of which allow selective labeling of cell surface thiols. Longer linkers should facilitate interaction between biotin and streptavidin. MTSEA biotin-XX MTSEA Biotincapcap; 2-((6-((6-((biotinoyl)amino)hexanoyl)amino)hexanoyl)amino)ethylmethanethiosulfonate Properties: White solid soluble in DMF or DMSO; Store desiccated at -20¿C; C25H45N5O6S3; MW: 607.7