Learn More
Biotium MTSEA Biotin-XX
Catalog No. NC0903451
Encompass_Preferred
MTSEA biotin selectively labels thiols under mild conditions. We offer MTSEA biotin with different linker lengths (Biotin-MTSEA, Biotin-X-MTSEA, and Biotin-XX-MTSEA), all of which allow selective labeling of cell surface thiols. Longer linkers should facilitate interaction between biotin and streptavidin. MTSEA biotin-XX: 2-((6-((6-((biotinoyl)amino)hexanoyl)amino)hexanoyl)amino)ethylmethanethiosulfonate; White solid soluble in DMF or DMSO; Store desiccated at -20°C; C25H45N5O6S3; MW: 607.7.
Catalog No. NC0903451 Supplier Biotium Supplier No. 900661
MTSEA biotin selectively labels thiols under mild conditions. We offer MTSEA biotin with different linker lengths (Biotin-MTSEA, Biotin-X-MTSEA, and Biotin-XX-MTSEA), all of which allow selective labeling of cell surface thiols. Longer linkers should facilitate interaction between biotin and streptavidin. MTSEA biotin-XX: 2-((6-((6-((biotinoyl)amino)hexanoyl)amino)hexanoyl)amino)ethylmethanethiosulfonate; White solid soluble in DMF or DMSO; Store desiccated at -20°C; C25H45N5O6S3; MW: 607.7.