Learn More
Medchemexpress LLC MTSEA hydrobromide | 16599-33-0 | 97.9% | 50 MG
SDP
Catalog No. 5000232195
Encompass_Preferred
MTSEA hydrobromide is a sulfhydryl-reactive reagent used to modify free cysteine residues in proteins, introducing a positively charged side chain approximately the size of lysine. The hydrobromide salt is supplied as a solid and is used in biochemical assays and site-directed cysteine modification to probe protein structure and function.
  • Sulfhydryl-reactive reagent for cysteine modification.
  • Introduces a positively charged side chain similar in size to lysine.
  • Suitable for biochemical assays and site-directed mutagenesis studies.
  • Soluble in water and common organic solvents such as DMSO.
  • High purity suitable for research applications.
Catalog No. 50-002-32195 Supplier Medchemexpress LLC Supplier No. HY12012850MG
MTSEA hydrobromide is a sulfhydryl-reactive reagent used to modify free cysteine residues in proteins, introducing a positively charged side chain approximately the size of lysine. The hydrobromide salt is supplied as a solid and is used in biochemical assays and site-directed cysteine modification to probe protein structure and function.
  • Sulfhydryl-reactive reagent for cysteine modification.
  • Introduces a positively charged side chain similar in size to lysine.
  • Suitable for biochemical assays and site-directed mutagenesis studies.
  • Soluble in water and common organic solvents such as DMSO.
  • High purity suitable for research applications.