Learn More
Biotium MTSES (FULL CHEMICAL NAME: SODIUM (2-SU
Supplier:  Biotium 91020
Encompass_Preferred
MTSES (2-Sulfonatoethyl methanethiosulfonate sodium salt) is a negatively charged MTS reagent. White solid soluble in DMSO Store desiccated at -20°C C3H7NaO5S3 MW: 242.21
Catalog No. NC0903429