Learn More
Biotium MTSET (FULL CHEMICAL NAME: [2-(TRIMETHY
Supplier:  Biotium 91021
Encompass_Preferred
MTSET (2-(Trimethylammonium)ethyl methanethiosulfonate, bromide) is a positively charged MTS reagent. Off-white solid soluble in water or DMSO Store desiccated at -20°C C6H16BrNO2S2 MW: 278.23
Catalog No. NC0903427