Learn More
American Radiolabeled Chemicals Inc MTSET 3H 50UC
SDP
Catalog No. 50901107
Encompass
MTSET-methyl-3H 50uCi in ethanol:water (1:1), 1 mCi/ml, S.A. 50-60 Ci/mmol, M.W. 278.24, Shipped in Dry Ice, Exclusive
Catalog No. 50-901-107 Supplier American Radiolabeled Chemicals Inc Supplier No. ART 176950UC
Add to Cart
Add to Cart
Request a Quote
MTSET-methyl-3H 50uCi in ethanol:water (1:1), 1 mCi/ml, S.A. 50-60 Ci/mmol, M.W. 278.24, Shipped in Dry Ice, Exclusive