Learn More
Cayman Chemical MTSETchlorIde 10mg
SDP
Catalog No. 502981596
Encompass_Preferred
A positively charged sulfhydryl-specific reagent that reacts with substituted-cysteines; used to provide functional information about relative positions of amino acids within a protein and to probe binding site electrostatic interactions
Catalog No. 50-298-1596 Supplier Cayman Chemical Supplier No. 2106910 MG
A positively charged sulfhydryl-specific reagent that reacts with substituted-cysteines; used to provide functional information about relative positions of amino acids within a protein and to probe binding site electrostatic interactions