Learn More
Enzo Life Sciences Matriptase (human), (recombinant) (His-tag) (250U)
SDP
Catalog No. 502007957
Encompass_Preferred
Alternative name: MT-SP1, Membrane type serine protease 1, ST14, Suppression of tumorigenicity protein 14, TADG-15, Tumor associated differentially expressed gene 15 protein, SNC19. Concentration: 100nM. Purity: ≥98% (SDS-PAGE). MW: ~25kDa. Formulation: Liquid. In 100mM TRIS, pH 8.0, containing 200mM sodium chloride, 1mM β-mercaptoethanol and 10% glycerol with a recommended assay buffer of 50mM HEPES, pH 7.4, 150mM NaCl, with 5mg/ml BSA. Source: Recombinant human matriptase (aa 596-855) fused at the N-terminus to a His-tag. Long Term Storage: -80°C. Use/Stability: Stable for at least 6 months when stored at -20°C. Do not store diluted solutions. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7957 Supplier Enzo Life Sciences Supplier No. ALX201246U250
Alternative name: MT-SP1, Membrane type serine protease 1, ST14, Suppression of tumorigenicity protein 14, TADG-15, Tumor associated differentially expressed gene 15 protein, SNC19. Concentration: 100nM. Purity: ≥98% (SDS-PAGE). MW: ~25kDa. Formulation: Liquid. In 100mM TRIS, pH 8.0, containing 200mM sodium chloride, 1mM β-mercaptoethanol and 10% glycerol with a recommended assay buffer of 50mM HEPES, pH 7.4, 150mM NaCl, with 5mg/ml BSA. Source: Recombinant human matriptase (aa 596-855) fused at the N-terminus to a His-tag. Long Term Storage: -80°C. Use/Stability: Stable for at least 6 months when stored at -20°C. Do not store diluted solutions. Handling: Avoid freeze/thaw cycles.