Learn More
AdipoGen MTSSL
SDP
Catalog No. 502562283
Encompass_Preferred
Chemical. CAS 81213-52-7. Formula C10H18NO3S2. MW 264.3. Highly reactive, thiol-specific spin-label. Specific conformational probe of thiol site structure by its minimal rotational freedom and distance from the covalent disulfide linkage to the macromolecule under study. Used to label cysteine residues in proteins site-directed labeling, SDS-labeling. Allows protein structure and protein dynamics determination as well as the study of protein-protein and protein-oligonucleotide interactions.
Catalog No. 50-256-2283 Supplier AdipoGen Supplier No. AGCR13573M050
Chemical. CAS 81213-52-7. Formula C10H18NO3S2. MW 264.3. Highly reactive, thiol-specific spin-label. Specific conformational probe of thiol site structure by its minimal rotational freedom and distance from the covalent disulfide linkage to the macromolecule under study. Used to label cysteine residues in proteins site-directed labeling, SDS-labeling. Allows protein structure and protein dynamics determination as well as the study of protein-protein and protein-oligonucleotide interactions.