Learn More
Abcam MTSSL, thiol-specific spin label, 10MG
Catalog No. 502705899
Encompass_Preferred
MW 264.4 Da, Purity >98%. Highly reactive thiol-specific spin label. For protein cysteine residue labeling, in protein structure and protein dynamics determination and in protein-protein and protein-oligonucleotide interaction studies. Active in vitro.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5899 Supplier Abcam Supplier No. AB14430810MG
MW 264.4 Da, Purity >98%. Highly reactive thiol-specific spin label. For protein cysteine residue labeling, in protein structure and protein dynamics determination and in protein-protein and protein-oligonucleotide interaction studies. Active in vitro.
The product is subject to the following: Abcam Restricted Use Statement