Learn More
Cayman Chemical MTT Cell Proliferation Assay Kit, 500 dtn
SDP
Catalog No. 502886885
Encompass_Preferred
Cayman’s MTT Proliferation Assay provides an easy to use tool for studying the induction and inhibition of cell proliferation in any in vitro model. This kit will also allow investigators to screen drug candidates involved in cell cycle regulation. In this assay, MTT is taken up by cells and then reduced to formazan by intracellular NAD(P)H-oxidoreductases.
Catalog No. 50-288-6885 Supplier Cayman Chemical Supplier No. 10009365500 DTN
Cayman’s MTT Proliferation Assay provides an easy to use tool for studying the induction and inhibition of cell proliferation in any in vitro model. This kit will also allow investigators to screen drug candidates involved in cell cycle regulation. In this assay, MTT is taken up by cells and then reduced to formazan by intracellular NAD(P)H-oxidoreductases.