Learn More
Apexbio Technology LLC MTT Cell Proliferation Assay Kit 2500T
SDP
Catalog No. 5000097323
Encompass_Preferred
MTT Cell Proliferation Assay Kit is a colorimetric assay designed to quantify cell viability proliferation or cytotoxicity in mammalian cell cultures by measuring metabolic activity The kit exerts its biological activity through the enzymatic reduction of the tetrazolium dye MTT by actively respiring mitochondria in viable cells leading to the formation of insoluble purple formazan crystals The amount of formazan produced correlates with cellular metabolic activity Based on these properties MTT Cell Proliferation Assay Kit holds research potential in pharmaceutical screening toxicity testing oncology and cell biology research
Catalog No. 50-000-97323 Supplier Apexbio Technology LLC Supplier No. K22492500
Add to Cart
Add to Cart
MTT Cell Proliferation Assay Kit is a colorimetric assay designed to quantify cell viability proliferation or cytotoxicity in mammalian cell cultures by measuring metabolic activity The kit exerts its biological activity through the enzymatic reduction of the tetrazolium dye MTT by actively respiring mitochondria in viable cells leading to the formation of insoluble purple formazan crystals The amount of formazan produced correlates with cellular metabolic activity Based on these properties MTT Cell Proliferation Assay Kit holds research potential in pharmaceutical screening toxicity testing oncology and cell biology research