Learn More
Cell Biolabs Inc MTT CELL PROLIFERATION ASSAY
Catalog No. 50672683
Encompass_Preferred
CytoSelect MTT Cell Proliferation Assay provides a colorimetric format for measuring cell proliferation. Reagents are sufficient for the evaluation of 960 assays in 96-well plates or 192 assays in 24-well plates.
Catalog No. 50-672-683 Supplier Cell Biolabs Inc Supplier No. CBA252
CytoSelect MTT Cell Proliferation Assay provides a colorimetric format for measuring cell proliferation. Reagents are sufficient for the evaluation of 960 assays in 96-well plates or 192 assays in 24-well plates.