Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MTT Formazan 98.0+%, TCI America™
SDP

Catalog No. B0275100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0275100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B0275100MG Supplier TCI America Supplier No. B0275100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 23305-68-2
Molecular Formula C18H17N5S
Molecular Weight (g/mol) 335.429
MDL Number MFCD00080731
InChI Key GIFKVQOPAJDJLO-RHRKZKCTSA-N
Synonym 1-(4,5-Dimethyl-2-thiazolyl)-3,5-diphenylformazan, 3,5-Diphenyl-1-(4,5-dimethyl-2-thiazolyl)formazan
PubChem CID 16218671
IUPAC Name N'-anilino-N-[(4,5-dimethyl-1,3-thiazol-2-yl)imino]benzenecarboximidamide
SMILES CC1=C(SC(=N1)N=NC(=NNC2=CC=CC=C2)C3=CC=CC=C3)C

Specifications

Melting Point 169°C
Color Black
Quantity 100 mg
Formula Weight 335.43
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material MTT Formazan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.