Learn More
Research Products International Corp MTT [Thiazolyl Blue Tetrazolium Bromide], 5 Grams
SDP
Catalog No. 50213524
Encompass_Preferred
Water soluble salt of Tetrazolium. Yields a yellowish solution when prepared in media or salt solutions lacking phenol red. By cleavage of the tetrazolium ring by dehydrogenase enzymes, dissolved MTT is converted into insoluble purple formazan.
Catalog No. 50-213-524 Supplier Research Products International Corp Supplier No. M920505.0
Water soluble salt of Tetrazolium. Yields a yellowish solution when prepared in media or salt solutions lacking phenol red. By cleavage of the tetrazolium ring by dehydrogenase enzymes, dissolved MTT is converted into insoluble purple formazan.