Learn More
Aviva Systems Biology Corp MTUS1 Antibody - N-terminal region (ARP44418_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44418_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against MTUS1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MTUS1. Alternate names: ATIP, DKFZp586D1519, DKFZp686F20243, FLJ14295, KIAA1288, MP44, MTSG1, ATBPApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58803