Learn More
Aviva Systems Biology Corp MTUS2 Antibody - C-terminal region (ARP62853_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62853_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MTUS2. It was validated on Western Blot. . Alternate names: CAZIP, KIAA0774, TIP150ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-786