Learn More
Medchemexpress LLC MtvkPabc-p5 TFA | 1190.22 | C54H74F3N11O16 | 5 MG
SDP
Catalog No. 5000204372
Encompass_Preferred
MTvkPABC-P5 TFA is a Toll-like receptor 7 (TLR7) agonist and immune stimulant used as a reagent for synthesizing immune-stimulating antibody conjugates (ISACs). It is supplied as the trifluoroacetic acid (TFA) salt in solid form. Molecular formula: C54H74F3N11O16; molecular weight: 1190.22.
  • Acts as a TLR7 agonist and immune stimulant.
  • Used to synthesize immune-stimulating antibody conjugates (ISACs).
  • Supplied as the trifluoroacetic acid (TFA) salt in solid form.
  • Storage: solid -20°C; protect from light; store under nitrogen.
  • Available pack sizes: 1 mg, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg.
Catalog No. 50-002-04372 Supplier Medchemexpress LLC Supplier No. HY156693A5MG
MTvkPABC-P5 TFA is a Toll-like receptor 7 (TLR7) agonist and immune stimulant used as a reagent for synthesizing immune-stimulating antibody conjugates (ISACs). It is supplied as the trifluoroacetic acid (TFA) salt in solid form. Molecular formula: C54H74F3N11O16; molecular weight: 1190.22.
  • Acts as a TLR7 agonist and immune stimulant.
  • Used to synthesize immune-stimulating antibody conjugates (ISACs).
  • Supplied as the trifluoroacetic acid (TFA) salt in solid form.
  • Storage: solid -20°C; protect from light; store under nitrogen.
  • Available pack sizes: 1 mg, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg.