Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mtvkpabc-p5 TFA | 98.8% | 1190.22 | C54H74F3N11O16 | 25 MG
SDP

Catalog No. 5000204419
Encompass_Preferred

MTvkPABC-P5 TFA is the trifluoroacetic acid (TFA) salt of a synthetic Toll-like receptor 7 (TLR7) agonist. It functions as an immune stimulant used in research applications, including the synthesis of immune-stimulating antibody conjugates (ISACs).

  • Acts as a TLR7 agonist and immune stimulant.
  • Suitable for synthesis of immune-stimulating antibody conjugates (ISACs).
  • Provided as a solid TFA salt.
  • High purity, 98.8%.
  • Molecular weight 1190.22, formula C54H74F3N11O16.
  • Available as 25 MG quantity.

Catalog No. 50-002-04419 Supplier Medchemexpress LLC Supplier No. HY156693A25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

MTvkPABC-P5 TFA is the trifluoroacetic acid (TFA) salt of a synthetic Toll-like receptor 7 (TLR7) agonist. It functions as an immune stimulant used in research applications, including the synthesis of immune-stimulating antibody conjugates (ISACs).

  • Acts as a TLR7 agonist and immune stimulant.
  • Suitable for synthesis of immune-stimulating antibody conjugates (ISACs).
  • Provided as a solid TFA salt.
  • High purity, 98.8%.
  • Molecular weight 1190.22, formula C54H74F3N11O16.
  • Available as 25 MG quantity.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.