Learn More
Medchemexpress LLC MTvkPABC-P5 TFA | 98.8% | 1190.22 Da | C54H74F3N11O16 | 1 MG
SDP
Catalog No. 5000204315
Encompass_Preferred
MTvkPABC-P5 TFA is the trifluoroacetic acid (TFA) salt form of a Toll-like receptor 7 (TLR7) agonist used as an immune stimulant and as a synthetic building block for immune-stimulating antibody conjugates (ISACs).
  • Acts as a Toll-like receptor 7 (TLR7) agonist and immune stimulant.
  • Used as a building block for synthesis of immune-stimulating antibody conjugates (ISACs).
  • Provided as a TFA salt for improved handling and stability.
  • Reported high purity suitable for research use.
  • Offered in small mg pack sizes for laboratory applications.
Catalog No. 50-002-04315 Supplier Medchemexpress LLC Supplier No. HY156693A1MG
MTvkPABC-P5 TFA is the trifluoroacetic acid (TFA) salt form of a Toll-like receptor 7 (TLR7) agonist used as an immune stimulant and as a synthetic building block for immune-stimulating antibody conjugates (ISACs).
  • Acts as a Toll-like receptor 7 (TLR7) agonist and immune stimulant.
  • Used as a building block for synthesis of immune-stimulating antibody conjugates (ISACs).
  • Provided as a TFA salt for improved handling and stability.
  • Reported high purity suitable for research use.
  • Offered in small mg pack sizes for laboratory applications.