Learn More
Medchemexpress LLC MTX-531 | 2791417-66-6 | 99.9% | 453.94 | 50 MG
SDP
Catalog No. 5000394951
Encompass_Preferred
MTX-531 is an oral drug that inhibits EGFR and PI3K, demonstrating anti-tumor effects. It also acts as a weak agonist of PPARγ, helping to alleviate hyperglycemia induced by PI3K inhibitors.
  • Inhibits epidermal growth factor receptor.
  • Inhibits phosphoinositide 3-kinase.
  • Demonstrates anti-tumor effects.
  • Acts as a weak agonist of peroxisome proliferator-activated receptors.
  • Helps alleviate hyperglycemia.
  • Suppresses tumor proliferation.
Catalog No. 50-003-94951 Supplier Medchemexpress LLC Supplier No. HY16271350MG
MTX-531 is an oral drug that inhibits EGFR and PI3K, demonstrating anti-tumor effects. It also acts as a weak agonist of PPARγ, helping to alleviate hyperglycemia induced by PI3K inhibitors.
  • Inhibits epidermal growth factor receptor.
  • Inhibits phosphoinositide 3-kinase.
  • Demonstrates anti-tumor effects.
  • Acts as a weak agonist of peroxisome proliferator-activated receptors.
  • Helps alleviate hyperglycemia.
  • Suppresses tumor proliferation.