Learn More
Cayman Chemical Methyl 5,6,7,8-Tetradehydro Risperidone, Purity: ≥98%, CAS: 106266-08-4, 10 mg
SDP
Catalog No. 502967419
Encompass_Preferred
An internal standard used in the quantitative analysis of risperidone
Catalog No. 50-296-7419 Supplier Cayman Chemical Supplier No. 1780110 MG
An internal standard used in the quantitative analysis of risperidone