Learn More
Medchemexpress LLC Mu-6S-Palm-β-Glc | 229644-17-1 | >99.9% | C32H48O8S | 1 MG
SDP
Catalog No. 5000354127
Encompass_Preferred
Mu-6S-Palm-β-Glc is a fluorogenic substrate for palmitoyl-protein thioesterase (PPT), primarily utilized in research contexts. This compound plays a role in the study of neuronal ceroid lipofuscin disease, particularly in infants.
  • Functions as a fluorogenic substrate for palmitoyl-protein thioesterase (PPT)
  • Employed in research for neuronal ceroid lipofuscin disease
  • Specifically aids in studies related to infant health
Catalog No. 50-003-54127 Supplier Medchemexpress LLC Supplier No. HY1455191MG
Mu-6S-Palm-β-Glc is a fluorogenic substrate for palmitoyl-protein thioesterase (PPT), primarily utilized in research contexts. This compound plays a role in the study of neuronal ceroid lipofuscin disease, particularly in infants.
  • Functions as a fluorogenic substrate for palmitoyl-protein thioesterase (PPT)
  • Employed in research for neuronal ceroid lipofuscin disease
  • Specifically aids in studies related to infant health