Learn More
Bon Opus Biosciences Recombinant Mouse Apolipoprotein E/ApoE (C-6His) 500ug
SDP
Supplier:  Bon Opus Biosciences CJ05500UG
Encompass_Preferred
Recombinant Mouse Apolipoprotein E is produced by our Mammalian expression system and the target gene encoding Glu19-Gln311 is expressed with a 6His tag at the C-terminus. Accession# P08226. Formulation: Lyophilized from a 0.2 μm filtered solution of 20mM PB,150mM NaCl,pH7.4. Bon Opus Cat# CJ05
Catalog No. 50-217-2809