Learn More
Bon Opus Biosciences Recombinant Mouse IL-2RB /CD122 (C-hFc) -500ug
SDP
Supplier:  Bon Opus Biosciences BP133500UG
Encompass_Preferred
Recombinant Mouse Interleukin-2 Receptor subunit beta is produced by our Mammalian expression system and the target gene encoding Ala27-Glu240 is expressed with a hFc tag at the C-terminus. Expression system: Human 293 Cells. Accession# P16297. Formulation: Lyophilized from a 0.2 um filtered solution of PBS, pH7.4. Bon Opus Cat# BP133
Catalog No. 50-208-0215