Learn More
Bon Opus Biosciences Recombinant Mouse FCRN Heterodimer -500ug
SDP
Supplier:  Bon Opus Biosciences BP181500UG
Encompass_Preferred
Recombinant Mouse IgG Fc fragment receptor transporter is produced by our Mammalian expression system and the target gene encoding Ser22-Ser297/Ile21-Met119 is expressed with a 6His tag at the C-terminus. Expression system: Human 293 Cells. Accession# Q61559/P01887. Formulation: Lyophilized from a 0.2 um filtered solution of PBS, pH 7.4. Bon Opus Cat# BP181
Catalog No. 50-208-0290