Learn More
Bon Opus Biosciences Recombinant Mouse FGF-4 -50ug
SDP
Catalog No. 502080399
Encompass_Preferred
Recombinant Mouse Fibroblast Growth Factor 4 is produced by our E.coli expression system and the target gene encoding Ser67­-Leu202 is expressed. Expression system: E.coli. Accession# P11403. Formulation: Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Bon Opus Cat# CR66
Catalog No. 50-208-0399 Supplier Bon Opus Biosciences Supplier No. CR6650UG
Recombinant Mouse Fibroblast Growth Factor 4 is produced by our E.coli expression system and the target gene encoding Ser67­-Leu202 is expressed. Expression system: E.coli. Accession# P11403. Formulation: Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Bon Opus Cat# CR66