Learn More
Bon Opus Biosciences Recombinant Mouse GPC3 (C-6His) -50ug
SDP
Supplier:  Bon Opus Biosciences C00R50UG
Encompass_Preferred
Recombinant Mouse Glypican-3 is produced by our Mammalian expression system and the target gene encoding Gln25-Met557 is expressed with a 6His tag at the C-terminus Expression system: Human Cells. Accession# Q8CFZ4. Formulation: Lyophilized from a 0.2 um filtered solution of PBS, 5% Trehalose, pH7.4. Bon Opus Cat# C00R
Catalog No. 50-208-0320