Learn More
Bon Opus Biosciences Recombinant Mouse HGF (C-6His) -50ug
SDP
Supplier:  Bon Opus Biosciences CC1350UG
Encompass_Preferred
Recombinant Mouse Hepatocyte growth factor is produced by our Mammalian expression system and the target gene encoding Gln33-Leu728 is expressed with a 6His tag at the C-terminus. Expression system: Human Cells. Accession# Q08048. Formulation: Lyophilized from a 0.2 um filtered solution of PBS, pH 7.4. Bon Opus Cat# CC13
Catalog No. 50-208-0380