Learn More
Bon Opus Biosciences Recombinant Mouse Interferon gamma/IFN-gamma -500ug
SDP
Supplier:  Bon Opus Biosciences BP118500UG
Encompass_Preferred
Recombinant Mouse Interferon gamma is produced by our Mammalian expression system and the target gene encoding His23-Cys155 is expressed. Expression system: Human 293 Cells. Accession# P01580. Formulation: Lyophilized from a 0.2 um filtered solution of PBS, pH7.4. Bon Opus Cat# BP118
Catalog No. 50-208-0191