Learn More
Bon Opus Biosciences Recombinant Mouse IFN gamma (C-His) -500ug
SDP
Supplier:  Bon Opus Biosciences BP047500UG
Encompass_Preferred
Recombinant Mouse IFN gamma is produced by Human 293 Cells. The target gene encoding H23-C155 is expressed with a 8His tag at the C terminus. Expression system: Human 293 Cells. Accession# P01580. Formulation: Lyophilized frum a 0.2 um filtered solution of PBS, pH7.4. Bon Opus Cat# BP047
Catalog No. 50-208-0124