Learn More
Bon Opus Biosciences Recombinant Mouse Interleukin-11/IL-11 (N-His) -500ug
SDP
Supplier:  Bon Opus Biosciences BP062500UG
Encompass_Preferred
Recombinant Mouse Interleukin-11 is produced by E.coli. The target gene encoding G23-L199 is expressed with a 6His tag at the N terminus. Expression system: E. coli. Accession# P47873. Formulation: Supplied as a 0.2 um filtered solution of 2umM Tris-HCl, 30umM NaCl, 10% Glycerol, pH8.0. Bon Opus Cat# BP062
Catalog No. 50-208-0137