Learn More
Bon Opus Biosciences Recombinant Mouse IL-1 Receptor Antagonist Protein/IL-1ra/IL-1RA 500ug
SDP
Supplier:  Bon Opus Biosciences CK20500UG
Encompass_Preferred
Recombinant Mouse Interleukin-1 Receptor Antagonist Protein is produced by our E.coli expression system and the target gene encoding Arg27-Gln178 is expressed. Accession# P25085. Formulation: Lyophilized from a 0.2 μm filtered solution of 20mM PB, 150mM NaCl, pH 7.4. Bon Opus Cat# CK20
Catalog No. 50-217-2817